Canine IL-1 beta ELISA VetSet
Catalog # KINFVS0130D-002
Supplier: Kingfisher Biotech
CAS Number:
Specifications
- VE:1 KIT
- Reaktivität der primären Antikörper:Dog
- Target protein:IL-1 beta
- Bezeichnung:Canine IL-1 beta ELISA VetSet, two plate kit
Specifications
About this item
The Canine IL-1 beta VetSet (ELISA development kit) is for the quantitative determination of Canine IL-1 beta in cell culture supernatants.
Kit contents: Canine IL-1 beta coated plate (2 plates), Canine IL-1 beta standard, Canine IL-1 beta detection antibody, Streptavidin-HRP.