Anion exchange HPLC columns, Dionex™, IonPac™ AS4A-SC
Supplier: DIONEX
Certificates
About this item
AS4A-SC columns are used to separate the inorganic anions in environmental water and are recommended for the fast analysis of inorganic anions in deverse matrices while using the carbonate eluents.
- Used in food beverages and other deverse sample matrices
- Compatible with sodium tetraborate gradient to optimise difficult separations
Specifications
- Catalog No:
- DION043175
- DION043174
- Max. Druck:
- 4000 psi
- 4000 psi
- pH-Bereich:
- 0 - 14
- 0 - 14