Anion exchange HPLC columns, Dionex™, IonPac™ AS14
: DIONEX
These columns are used to separate the inorganic anions from the deverse matrices.
- Catalog No:
- DION046134
- DION046124
- Max. Druck:
- 4000 psi
- 4000 psi
- pH-Bereich:
- 2 - 12
- 2 - 12
Frequently Bought Together