Microtube racks, reversible, with removable handles
Catalog # DELTM-563A
Supplier: DELTALAB
Specifications
- Für Röhrchen Ø:12/8 mm
- Form:Reversible
- Anz. Bohrungen:80/60
- Farbe:Blau
- VE:10
Specifications
About this item
PP, reversible.
- Stackable
- By removing the handles, units can be firmly anchored laterally to one another
The racks have two sides for different tubes: one side holds up to 80 tubes of 12 mm diameter (for example 10×75 mm or 12×75 mm tubes, microtubes, etc.) and the other side holds up to 60 microtubes, PCR, etc.