Anion exchange HPLC column, Dionex™, IonPac™ AS22
Supplier: DIONEX
Certificates
About this item
Columns are designed for the fast analysis of inorganic anions in drinking water, ground water and waste water, by using isocratic carbonate/bicarbonate eluent common inorganic also can be separated easily.
Specifications
- Catalog No:
- DION064141
- DION064139
- Max. Druck:
- 3000 psi
- 3000 psi
- pH-Bereich:
- 0 - 14
- 0 - 14