Anion exchange HPLC columns, Dionex™, IonPac™ AS11
Anbieter: DIONEX
Zertifikate
Über diesen Artikel
IonPac AS11 column designed for fast gradient screening inorganic anions and organic acid anions in sample matrices such as foods, beverages, chemical process solutions, wastewaters, brines and power plant waters.
- Ideal for highly charged anions including polyphosphates and polycarboxylates
- Short analysis time and speeds throughput
Technische Daten
- Catalog No:
- DION044076
- DION044078
- Max. Druck:
- 4000 psi
- 4000 psi
- pH-Bereich:
- 0 - 14
- 0 - 14