Counting chamber with grid
Counting chambers with grid, 20×20 mm
Katalognr. DIVEGRID-1000
Anbieter: Diversified Biotech
Direkt zu
Product Details & Documents
Disposable polyester with acrylic adhesive grid stickers for micro or semi-micro objects.
- Inert and stable under most conditions
- Compatible with different kinds of microscopes
- Separate coverslips can be placed over each sticker
These slide grids are either used with dry or suspended specimens, such as cells held in buffer, under cover slips.
Technische Daten
- B×T:20×20 mm
- Bezeichnung:Counting chambers with grid