Combi top racks
Anbieter: KOEHLER TECHNISCHE PRODUKTEN
Direkt zu
Über diesen Artikel
Racks, made of polypropylene, neon coloured racks have four sides that can be used individually. with temperature resistant from −80 to 121 °C.
- Temperature resistant from −80 to 121 °C.
- Excellent chemical resistance
- Sturdy
2 Varianten
Product Details & Documents
Racks, made of polypropylene, neon coloured racks have four sides that can be used individually. with temperature resistant from −80 to 121 °C.
- Temperature resistant from −80 to 121 °C.
- Excellent chemical resistance
- Sturdy
Product Family Options
Produktinformationen