Dialysis bag clips, SP Bel-Art
Catalog # 229-0003
Supplier: Bel-Art Products, a Part of SP
Specifications
- Farbe:Durchscheinend
- Bezeichnung:Dialysis bag holder, for fitting onto the side of an open container, beaker or pitcher up to 1⁄4" thick
- Material:PP
- VE:6
Specifications
About this item
These strong clips made from PP are ideal for suspending dialysis bags in large beakers or pitchers up to 0,63 cm (¼") thick.
- Bags slide easily into the slot and then the holder can be clipped onto the container’s edge
- Smooth PP surface may be marked with identifying sequence numbers
- Autoclavable