Separating funnel stands, SP Bel-Art
Katalognr. BELAH189640000
Anbieter: Bel-Art Products, a Part of SP
Technische Daten
- Für:6 separating funnels of different sizes, 530×350 mm
- Environmentally Preferable:
- VE:1
Technische Daten
Über diesen Artikel
This rotary separatory funnel rack consists of a rectangular base, screw-in column and a support disc held in place by a screw clamp.
- Support disc is fully adjustable for different heights up to 21"
- Open front type support holes for easy removal and placement of separatory funnels
- Shipped unassembled
Ordering information: Includes 18 insert clips to accommodate different sized funnels.