Order Entry
Switzerland
ContactUsLinkComponent
(R)-(-)-2-Fluor-α-methyl-4-biphenylessigsäure
(R)-(-)-2-Fluor-α-methyl-4-biphenylessigsäure
Catalog # MOLE83025196-500MG
Supplier:  Molekula
CAS Number:  
(R)-(-)-2-Fluor-α-methyl-4-biphenylessigsäure
Catalog # MOLE83025196-500MG
Supplier:  Molekula
CAS Number:  

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :244.260 g/mol
EINECS:257-264-7
CAS Number:51543-40-9
Formula:C₁₅H₁₃FO₂
MDL Number:MFCD00869714