Pipette controller, Pipet-Aid® Hood Mate®
Katalognr. DRUM4-000-302
Anbieter: DRUMMOND
Technische Daten
- Kapazität:1 - 100 ml
- Bezeichnung:Pipette controller, Pipet-Aid® Hood Mate®, with EU-plug
- VE:1
Technische Daten
Über diesen Artikel
This controller is specifically designed for convenient, comfortable pipetting under a hood. Ideal for transfering liquids with the use of serological, volumetric and/or blow-out pipettes in either glass, plastic or something similar.
- Longer coiled hose
- Lightweight and manufactured from UV resistant materials
- Extremely lightweight handle provides nine fill and empty speed settings and proportional fingertip control
- Handle holder incorporates 'auto-off' feature: automatically shuts unit off after 3 m of non-use
- Dedicated pump housing attaches to hood wall which saves space and prevents spills
- Coiled vacuum/pressure line entering handle from the top prevents accidental knocking over of bench items