Adsorbents for column chromatography, Florisil®
Catalog # MANA815710.1
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Partikelgröße:150/250 µm
- VE:1 kg
Specifications
About this item
Hard granular magnesia silica gel: MgO (15,5 ±0,5%) SiO2 (84,0 ±0,5%) Na2SO4 (≤1,0%) 60/100 mesh.
Typical application: sample preparation (SPE); clean-up of pesticide residues, separation of chlorinated pesticides, extraction of steroids, sex hormones, antibiotics, lipids etc.