Order Entry
Switzerland
ContactUsLinkComponent
Ethanol ≥70% (v/v)
Catalog # VWRS83954.440
Supplier:  BDH Prolabo
CAS Number:  
Ethanol ≥70% (v/v)
Catalog # VWRS83954.440
Supplier:  BDH Prolabo
CAS Number:  

Specifications

  • Packung:
    Kunststoffkanister
  • VE:
    20 L
  • Seq:
    5302

Specifications

About this item

 :
 :46.07 g/mol
Storage Temperature:Raumtemperatur
EINECS:200-578-6
UN:1170
ADR:3,III
 :GHS02
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568