Protective apron, 9330 LTHR
KWIN100867240
- Anwendungsbereich Outfit:Schweißen
- Geschlecht:Unisex
- Farbe:Braun
- L×B:870×560 mm
- Modell:9330
- Stoff:Leather/nylon
- VE:1
Split leather apron with bib.
- Nylon waist and neck straps
Suitable for use in welding applications.