1-Boc-2-indolboronsäure
Marketsource
Supplier: ACTIVATE SCIENTIFIC
Jump to:
About this item
Synonyms: N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acid, 1H-Indole-2-boronic acid, N-BOC protected, N-Boc-indole-2-boronic Acid, 1-(tert-Butoxycarbonyl)indole-2-boronic acid, N-Boc-indole-2-boronic acid
- CAS Number:213318-44-6
- Formula:C₁₃H₁₆BNO₄
- MDL Number:MFCD02093045
- Molecular Weight:261.08 g/mol
2 Options Available Below
Product Details & Documents
Synonyms: N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acid, 1H-Indole-2-boronic acid, N-BOC protected, N-Boc-indole-2-boronic Acid, 1-(tert-Butoxycarbonyl)indole-2-boronic acid, N-Boc-indole-2-boronic acid
- CAS Number:213318-44-6
- Formula:C₁₃H₁₆BNO₄
- MDL Number:MFCD02093045
- Molecular Weight:261.08 g/mol
Product Family Options
Product Information
- PackungVEAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



