Bromphenolblau
Glass bottle for solids, 10 g
Katalognr. NADIHS-603-10G
Marketsource
Anbieter: NATIONAL DIAGNOSTICS
Bromphenolblau
Katalognr. NADIHS-603-10G
Anbieter: NATIONAL DIAGNOSTICS
Marketsource
CAS Nummer: 115-39-9
Direkt zu
Product Details & Documents
Synonyme: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
- CAS Nummer:115-39-9
- Formel:C₁₉H₁₀Br₄O₅S
- MDL-Nummer:MFCD00005875
- Molekulargewicht:669.97 g/mol
- Merck-Index:13,01429
- Schmelzender Platingehalt:273 °C
- Dichte:1 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:204-086-2
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:10 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



