Okadasäure
Supplier: LATOXAN
Jump to:
About this item
Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin, OA
- CAS Number:78111-17-8
- Formula:C₄₄H₆₈O₁₃
- MDL Number:MFCD00083455
- Molecular Weight:805.02 g/mol
2 Options Available Below
Product Details & Documents
Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin, OA
- CAS Number:78111-17-8
- Formula:C₄₄H₆₈O₁₃
- MDL Number:MFCD00083455
- Molecular Weight:805.02 g/mol
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



