m-FC with rosolic acid broth
Katalognr. HACH2428520
Anbieter: Hach
CAS-Nummer:
Technische Daten
- VE:20
- Bezeichnung:m-FC with rosolic acid broth, 2 ml glass ampule
- Für:Faecal Coliforms
Technische Daten
Über diesen Artikel
Selectivity: Faecal coliform bacteria. For drinking water, wastewater, ambient water, food, and beverage applications.
- Save valuable time and money using this prepared media for membrane filtration methods
- Procedures, prepared media, and required consumables available for presumptive and confirming tests