Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Peppermint oil
Pfefferminzöl
Pfefferminzöl
Pfefferminzöl
Catalog # ROTH3329.1
Supplier:
Roth Carl
CAS Number:
Expand image
Pfefferminzöl
Catalog # ROTH3329.1
Supplier:
Roth Carl
CAS Number:
Specifications
Packung:
Glass bottle
VE:
500 mL
Seq:
2725
Specifications
About this item
Product Details
:
CAS Number:
8006-90-4
MDL Number:
MFCD00147870
Product Details