Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
4-Hydroxy-2,6-dimethoxybenzaldehyd

4-Hydroxy-2,6-dimethoxybenzaldehyd

Marketsource
Supplier:   ACTIVATE SCIENTIFIC
4-Hydroxy-2,6-dimethoxybenzaldehyd
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  2,6-Dimethoxy-4-hydroxybenzaldehyde
  • CAS Number:22080-96-2
  • Formula:C₉H₁₀O₄
  • MDL Number:MFCD01407645
  • Molecular Weight:182.1733 g/mol
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Packung
    VE
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ACTIAS35045G1
    Marketsource
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ACTIAS35045G5
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.