Formaldehyd 5% in wässriger Lösung, VOLUSOL®, gepuffert
VWRS6170.5000
:
- Packung:Kunststoffkanister
- VE:5 L
- Seq:5190
- Environmentally Preferable:
: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
:
:30.03 g/mol
:200-001-8
:GHS08
:GHS07
:Raumtemperatur
:50-00-0
:CH₂O
:MFCD00003274