HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)
Catalog # RC-121
Supplier: G-Biosciences
CAS Number:
Specifications
- Packung:Glass bottle for solids
- VE:250 g
- Seq:3545
Specifications
About this item
:
Merck Index:13,04672
Melting Pt:210…215 °C
Density:1.448 g/cm³ (20 °C)
:238.31 g/mol
Storage Temperature:Raumtemperatur
EINECS:230-907-9
CAS Number:7365-45-9
Formula:C₈H₁₈N₂O₄S
MDL Number:MFCD00006158