Trypanblau
Anbieter: EUROCLONE
Direkt zu
Über diesen Artikel
Synonyme: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
- CAS Nummer:72-57-1
- Formel:C₃₄H₂₄N₆Na₄O₁₄S₄
- MDL-Nummer:MFCD00003969
- Molekulargewicht:960.82 g/mol
- EINECS:200-786-7
3 Varianten
Product Details & Documents
Synonyme: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
- CAS Nummer:72-57-1
- Formel:C₃₄H₂₄N₆Na₄O₁₄S₄
- MDL-Nummer:MFCD00003969
- Molekulargewicht:960.82 g/mol
- EINECS:200-786-7
Product Family Options
Produktinformationen
- PackungVEVerfügbarkeitPreis



