ContactUsLinkComponent
Stands with funnel holder
Catalog # VITL78294
Supplier:  VITLAB
Stands with funnel holder
Catalog # VITL78294
Supplier:  VITLAB
Specifications
  • Bezeichnung:
    Stand with funnel holder for four funnels
  • L×B:
    450×140 mm
  • Seq:
    02
  • VE:
    1
Specifications
About this item
Funnel holder with base, made from PP. Support rod made from aluminium.
  • For two or four funnels with an max. outer diameter of 50 to 120 mm
  • Adjustable height