ContactUsLinkComponent
Laboratory trays
Laboratory trays
  VITL165094
 :  VITLAB
undefined
Laboratory trays
  VITL165094
 :  VITLAB
  • Farbe:
    Weiß
  • Seq:
    01
  • B×T×H:
    230×180x42 mm
  • VE:
    1
Laboratory trays, made of white PP.
  • Robust design
  • Easy to clean
  • Good chemical resistance
  • Smooth surface, rounded corners and edges
Ideal for a wide range of applications.