Tubing connectors, Y-shaped
VITL80468
: VITLAB
- Schlauch Innen-Ø:12 mm
- Material:PP
- Seq:05
- VE:20
Connectors, Y-shaped, made of PP, for the splitting or combining of liquid lines in a tubing system.
- Good mechanical strength and chemical resistance
- Excellent thermal resistance
- Autoclavable
Dimensions are shown as both nominal and exact as elasticity of tubing will influence the choice of size.