3-Chlorisochinolin
Marketsource
Anbieter: ACTIVATE SCIENTIFIC
Direkt zu
Über diesen Artikel
- CAS Nummer:19493-45-9
- Formel:C₉H₆ClN
- MDL-Nummer:MFCD05982009
- Molekulargewicht:163.6000 g/mol
2 Varianten
Product Details & Documents
- CAS Nummer:19493-45-9
- Formel:C₉H₆ClN
- MDL-Nummer:MFCD05982009
- Molekulargewicht:163.6000 g/mol
Product Family Options
Produktinformationen
- PackungVEVerfügbarkeitPreis
- Spezifikationen:MarketsourceMarketsource
- Spezifikationen:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



