Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
(R)-(-)-Mandelsäure

(R)-(-)-Mandelsäure

Glass bottle for solids, 100 g
Catalog # ACTIAS20123G100
Corrosive
Marketsource
Supplier:   ACTIVATE SCIENTIFIC
(R)-(-)-Mandelsäure
Catalog # ACTIAS20123G100
Marketsource
CAS Number:  611-71-2
(R)-(-)-Mandelsäure

Glass bottle for solids, 100 g

CAS Number:  611-71-2
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
  • CAS Number:611-71-2
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00064251
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:322 °C (1013 hPa)
  • Melting Pt:130…132 °C
  • Density:1,341 g/cm³ (20 °C)
  • Flash Pt:>190 °C
  • EINECS:210-276-6
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.