$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
INITIAL LOAD
CA10019-186
:
Biolegend
New Product
INITIAL LOAD
CA10019-186
:
Biolegend
:
149502
New Product
Frequently Bought Together
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Light-Duty Tissue Wipers
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Universal Pipette Tips
: VWR International
Acétonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma