N-(2-HYDROXYETHYL)-N-METHYLPERFLUORO 1ML
Catalogue # SXCRM109791150
Fournisseur: SPEX CERTIPREP LLC
New Product
N-(2-HYDROXYETHYL)-N-METHYLPERFLUORO 1ML
Catalogue # SXCRM109791150
Fournisseur: SPEX CERTIPREP LLC
Supplier Number: CRM10979.11-50-ME
New Product
Frequently Bought Together
AZOTOBACTER BROTH MANNITOL500G
Fournisseur: HiMedia
VACUETTE® Serum Clot Activator Tubes, Greiner Bio-One
Fournisseur: Greiner Bio-One
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Fournisseur: Air-Tite
Calcium chloride dihydrate 99.0-107.0% USP, Ph. Eur., BP, JP endotoxin tested
Fournisseur: Avantor Performance Materials
Acétone ACS for production, VWR Chemicals BDH®
Fournisseur: VWR International
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Fournisseur: MEDONE SURGICAL, INC
VWR® Specimen Containers, PP
Fournisseur: VWR International
Ocean Pacific® Ninja™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Fournisseur: Medicom
Diethyl phthalate ≥99.0%
Fournisseur: TCI America
DIBMA Screening Kit, HEPES
Fournisseur: CUBE BIOTECH
Potassium hexafluorophosphate ≥99%
Fournisseur: Thermo Scientific Chemicals
Anti-TH Mouse Monoclonal Antibody [clone: 2/40/15]
Fournisseur: Biolegend
Acide acétique glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Fournisseur: Hardy Diagnostics
Toluene ACS
Fournisseur: INNOVA CHEMICALS CA
Calcium chloride, dried 97%, powder
Fournisseur: Thermo Scientific Chemicals
EASY III™ Rapid Differential Stains, Azer Scientific
Fournisseur: Azer Scientific Inc.