EDB DBP TCP DRINK H2O PT EXPRESS PK2
Catalogue # SXQCO-007-4B
Fournisseur: SPEX CERTIPREP LLC
New Product
EDB DBP TCP DRINK H2O PT EXPRESS PK2
Catalogue # SXQCO-007-4B
Fournisseur: SPEX CERTIPREP LLC
Supplier Number: QCO-007-4B
New Product
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Mineral oil, white (petroleum) light, high purity
Fournisseur: VWR
Ethyl Alcohol Denatured
Fournisseur: Ward's Science
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Fournisseur: Air-Tite
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
Fournisseur: VWR International
VWR® Sterile 70% Isopropanol
Fournisseur: VWR International
Electrode Cleaning and Storage Solutions, Oakton®, Cole Parmer
Fournisseur: Antyila Scientific
BD Syringes only with Luer-Lok™, Slip or Eccentric tips
Fournisseur: BD
Tryptone to 2xYT Broth, Life Technologies
Fournisseur: BD
VWR® Specimen Containers, PP with PE Caps
Fournisseur: VWR International
Mounting Medium, Epredia™
Fournisseur: Thermo Scientific
Cetrimide Select Agar, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA
Toluène ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
CL17 DPD Indicator, Hach
Fournisseur: Hach
MORPHOLINE STANDARD, 1000PPM
Fournisseur: Labchem
InLab® Conductivity Probes, Mettler Toledo
Fournisseur: Mettler Toledo
Giemsa Stain
Fournisseur: Ward's Science
Toluene ACS
Fournisseur: INNOVA CHEMICALS CA