Sun, Earth, Moon Systems Model | Avantor
Sun, Earth, Moon Systems Model
Replacement light bulb
Catalog # 470359-426
New Product
Supplier:   Cochranes of Oxford
Fournisseur #:  1075
Sun, Earth, Moon Systems Model
Catalog # 470359-426
New Product
Sun, Earth, Moon Systems Model
Sun, Earth, Moon Systems Model
Replacement light bulb
Price
$9.40
Log in pour la disponibilité
Each
Product Details & Documents
Simulates Seasons, Phases of the Moon and More
  • Demonstrates solar and lunar eclipses
  • Contains two sizes of Earth - Moon setups
Demonstrate night and day, the seasons, phases of the moon, and eclipses with this robust and flexible model. More accurate demonstrations are made possible with two different sizes of Earth and Moon setups, and geared manual rotation system. In the large–scale setup, a powerful light casts visible shadows onto the 4 " Earth. The smaller version demonstrates the phases of the moon and frequency of solar and lunar eclipses as the moon rotates on an inclined orbit. Power: 120 VAC. Size: 25L×9⁷/₈".
Ordering information: Includes: 1 sun, earth, moon systems model base
1 Small earth model
1 Large earth model
1 Small moon Model
1 Large moon Model
1 Model person
1 Date, eclipse, and phase of the moon indicator card set
1 Teacher guide
Spécifications
  • Désignation:
    Replacement light bulb
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.