Sun, Earth, Moon Systems Model
Product Details & Documents
Simulates Seasons, Phases of the Moon and More
- Demonstrates solar and lunar eclipses
- Contains two sizes of Earth - Moon setups
Demonstrate night and day, the seasons, phases of the moon, and eclipses with this robust and flexible model. More accurate demonstrations are made possible with two different sizes of Earth and Moon setups, and geared manual rotation system. In the large–scale setup, a powerful light casts visible shadows onto the 4 " Earth. The smaller version demonstrates the phases of the moon and frequency of solar and lunar eclipses as the moon rotates on an inclined orbit. Power: 120 VAC. Size: 25L×9⁷/₈".
Ordering information: Includes: 1 sun, earth, moon systems model base
1 Small earth model
1 Large earth model
1 Small moon Model
1 Large moon Model
1 Model person
1 Date, eclipse, and phase of the moon indicator card set
1 Teacher guide
Spécifications
- Désignation:Replacement light bulb