Avancé
SDS
Certificats
Données du fabricant
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
3/8" Y ASSEMBLY W/ 1-1/2" TC'S (X3)
Catalog # 1010747.
Supplier:
AVANTOR FLUID HANDLING, LLC
New Product
Expand image
3/8" Y ASSEMBLY W/ 1-1/2" TC'S (X3)
Catalog # 1010747.
Supplier:
AVANTOR FLUID HANDLING, LLC
Supplier Number:
1010747
New Product
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH