Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Cell Strainers, Sterile | Avantor

Cell Strainers, Sterile

Cell strainer, 100 μm, Sterile, Yellow, Polypropylene, Individually wrapped
Catalog # 77972-876
New Product
Fournisseur #:  229485
Cell Strainers, Sterile
Catalog # 77972-876
New Product
Cell Strainers, Sterile
Cell Strainers, Sterile

Cell strainer, 100 μm, Sterile, Yellow, Polypropylene, Individually wrapped

Price
$264.41
Log in pour la disponibilité
Case of 50
Product Details & Documents

Cell strainers for isolating single cells and removing debris or aggregate from cell suspensions.

  • Can be used for many applications including isolating cells and removing clumps or debris
    Hydrophilic treatment increases flow and reduces the effects of surface tension
    Designed to universally fit most brands of 50 ml conical tubes
    Extended lip on housing enables aseptic handling with forceps
Spécifications
  • Sterility:
    Sterile
  • Color:
    Yellow
  • Désignation:
    Cell strainer, 100 μm
  • Matière:
    Polypropylene
  • Packaging:
    Individually wrapped