Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Cell Strainers, Sterile | Avantor

Cell Strainers, Sterile

Cell strainer, 70 μm, Sterile, White, Polypropylene, Individually wrapped
Catalog # 77972-872
New Product
Fournisseur #:  229483
Cell Strainers, Sterile
Catalog # 77972-872
New Product
Cell Strainers, Sterile
Cell Strainers, Sterile

Cell strainer, 70 μm, Sterile, White, Polypropylene, Individually wrapped

Price
264,41 $
Log in pour la disponibilité
Case of 50
Product Details & Documents

Cell strainers for isolating single cells and removing debris or aggregate from cell suspensions.

  • Can be used for many applications including isolating cells and removing clumps or debris
    Hydrophilic treatment increases flow and reduces the effects of surface tension
    Designed to universally fit most brands of 50 ml conical tubes
    Extended lip on housing enables aseptic handling with forceps
Spécifications
  • Sterility:
    Sterile
  • Color:
    White
  • Désignation:
    Cell strainer, 70 μm
  • Matière:
    Polypropylene
  • Packaging:
    Individually wrapped
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.