Cell Strainers, Sterile
Cell Strainers, Sterile
Pestle, cell strainer, Sterile, Green, Polypropylene, Individually wrapped
Catalog # 77972-866
New Product
Supplier #:  229480
Cell Strainers, Sterile
Catalog # 77972-866
New Product
Cell Strainers, Sterile
Cell Strainers, Sterile
Pestle, cell strainer, Sterile, Green, Polypropylene, Individually wrapped
Price
$114.54
Log in for availability
Case of 50
Product Details & Documents
Cell strainers for isolating single cells and removing debris or aggregate from cell suspensions.
  • Can be used for many applications including isolating cells and removing clumps or debris
    Hydrophilic treatment increases flow and reduces the effects of surface tension
    Designed to universally fit most brands of 50 ml conical tubes
    Extended lip on housing enables aseptic handling with forceps
Specifications
  • Sterility:
    Sterile
  • Color:
    Green
  • Désignation:
    Pestle, cell strainer
  • Matière:
    Polypropylene
  • Packaging:
    Individually wrapped