Plasteur® Pasteur Pipets
Bulk packed, sterile, 9"
Catalog # 77972-594
New Product
Supplier: CELLTREAT Scientific Products
Fournisseur #: 229286
Jump to:
Product Details & Documents
Plasteur pipets for transferring liquids.
- CELLTREAT® exclusive product
Polypropylene is autoclavable and chemical resistant and provides chemical resistivity needed for many applications
Flexible plastic helps prevent glove or skin punctures
Available packaged sterile individually wrapped or bulk packed
Available packaged non-sterile in re-sealable bags or tab lock boxes
Eliminate sharps disposal costs
Spécifications
- Désignation:Bulk packed, sterile
- Longueur:9"