Plasteur® Pasteur Pipets | Avantor
Plasteur® Pasteur Pipets
Bulk packed, sterile, 9"
Catalog # 77972-592
New Product
Fournisseur #:  229285
Plasteur® Pasteur Pipets
Catalog # 77972-592
New Product
Plasteur® Pasteur Pipets
Plasteur® Pasteur Pipets
Bulk packed, sterile, 9"
Price
$374.46
Log in pour la disponibilité
Case of 500
Product Details & Documents
Plasteur pipets for transferring liquids.
  • CELLTREAT® exclusive product
    Polypropylene is autoclavable and chemical resistant and provides chemical resistivity needed for many applications
    Flexible plastic helps prevent glove or skin punctures
    Available packaged sterile individually wrapped or bulk packed
    Available packaged non-sterile in re-sealable bags or tab lock boxes
    Eliminate sharps disposal costs
Spécifications
  • Désignation:
    Bulk packed, sterile
  • Longueur:
    9"