Plasteur® Pasteur Pipets | Avantor
Plasteur® Pasteur Pipets
Individually wrapped, sterile, 9"
Catalog # 77972-586
New Product
Fournisseur #:  229280
Plasteur® Pasteur Pipets
Catalog # 77972-586
New Product
Plasteur® Pasteur Pipets
Plasteur® Pasteur Pipets
Individually wrapped, sterile, 9"
Price
$195.92
Log in pour la disponibilité
Case of 200
Product Details & Documents
Plasteur pipets for transferring liquids.
  • CELLTREAT® exclusive product
    Polypropylene is autoclavable and chemical resistant and provides chemical resistivity needed for many applications
    Flexible plastic helps prevent glove or skin punctures
    Available packaged sterile individually wrapped or bulk packed
    Available packaged non-sterile in re-sealable bags or tab lock boxes
    Eliminate sharps disposal costs
Spécifications
  • Désignation:
    Individually wrapped, sterile
  • Longueur:
    9"