BrainStorm STEM Earthquake Simulator Kit
Product Details & Documents
Simulate seismic activity to explore structural integrity and earthquake science.
Designed by teachers, for teachers
- Demonstrates structural design and stability
- Perfect for home or the classroom
- Available as a single kit or a class pack of ten with educator guide
- No glue or mess
The BrainStorm earthquake simulator STEM kit helps students investigate structural integrity through realistic seismic testing. Learners build their own structures and test them on a vibrating tremor table to understand how buildings respond to different frequencies and intensities of movement. This activity enhances STEM concepts in physics and engineering. These kits are authenticated as educational kits by STEM.org.
Delivery information: Kit includes components for structure and tremor table, assembly instructions, and educator support materials.
Spécifications
- Désignation:BrainStorm earthquake simulator STEM kit, single