Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Linsitinib ≥98%

50 mg
Catalog # ABCA_AB273391-50MG
New Product
Supplier:   Abcam
 :  AB273391-50MG
Linsitinib ≥98%
Catalog # ABCA_AB273391-50MG
Supplier:  Abcam
New Product
CAS Number:  867160-71-2
Linsitinib ≥98%

50 mg

CAS Number:  867160-71-2
Price
$1,450.00
Log in pour la disponibilité
Each (50mg)
 
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Potent and selective dual IGF-1R (IC₅₀ = 35 nM in cell-free assay) and Insulin Receptor (IC₅₀ = 75 nM in cell-free assay) kinase inhibitor. Inactive towards Abl, ALK, BTK, EGFR, FGFR1/2 and PKA. Displayed <50% inhibition at 1 μM against a panel of 88 kinases. Blocks IGF-1R autophosphorylation and downstream activation of Akt, ERK1/2 and S6 kinase. Blocks tumor cell line proliferation. Blocks IGF-1R driven tumor xenograft growth.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymes:  Cis-3-(8-Amino-1-(2-phenylquinolin-7-yl)imidazo[1,5-a]pyrazin-3-yl)-1-methylcyclobutan-1-ol
  • CAS Number:867160-71-2
  • Molecular Weight:421.5 g/mol
Spécifications

Résultats des tests de spécification

  • Constituents::
    98% 3-[8-amino-1-(2-phenylquinolin-7-yl)imidazo[1,5-a]pyrazin-3-yl]-1-methylcyclobutan-1-ol

Spécifications

  • Taille:
    50 mg

Spécifications