Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam | Avantor

Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam

Tissue Clearing Permeabilization Buffer - hydrophobic, 100 ml
Catalog # ABCA_AB243307-100M
New Product
Supplier:   Abcam
Fournisseur #:  AB243307-100ML
Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam
Catalog # ABCA_AB243307-100M
Supplier:  Abcam
New Product
CAS Number:  
Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam

Tissue Clearing Permeabilization Buffer - hydrophobic, 100 ml

CAS Number:  
Price
$359.00
Log in pour la disponibilité
Each (100ml)
La quantité minimale de commande est de 1
Product Details & Documents

Tissue Clearing Permeabilization Buffer ab243307 is part of our range of tissue clearing reagents and products that enable you to set up tissue clearing easily and quickly.

  • Constituents: 94% Water, 6% Sodium lauryl sulfate

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Spécifications

Spécifications

  • Désignation:
    Tissue Clearing Permeabilization Buffer - hydrophobic
  • Taille:
    100 ml
  • Cat. No.:
    ABCA_AB243307-100M
  • Storage temperature:
    Store at Ambient.

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.