Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Extraction Enhancer Buffer 50X, Abcam

Extraction Enhancer Buffer 50X, 1 ml
Catalog # ABCA_AB193971-1ML
New Product
Supplier:   Abcam
 :  AB193971-1ML
Extraction Enhancer Buffer 50X, Abcam
Catalog # ABCA_AB193971-1ML
Supplier:  Abcam
New Product
CAS Number:  
Extraction Enhancer Buffer 50X, Abcam

Extraction Enhancer Buffer 50X, 1 ml

CAS Number:  
Price
$91.60
Log in for availability
Each (1ml)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Abcam's Extraction Enhancer Buffer 50X is formulated for the preparation of cell or tissues extracts for use in immunoassays.

 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Désignation:
    Extraction Enhancer Buffer 50X
  • Taille:
    1 ml
  • Cat. No.:
    ABCA_AB193971-1ML
  • Storage temperature:
    Store at +4°C.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.