BARCODING PACK ADV TRAINING PP2-011325
Catalogue # MP77778-302
Fournisseur: QUANTUM-SI
New Product
BARCODING PACK ADV TRAINING PP2-011325
Catalogue # MP77778-302
Fournisseur: QUANTUM-SI
Supplier Number: 700-00004-00
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Fournisseur: BDH
Sodium hydroxyde 0.5 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
VWR® Traceable® Conductivity Calibration Standards (CRM)
Fournisseur: VWR International
Formalin 10%, neutral buffered, VWR®
Fournisseur: VWR International
Cytoslide™ Microscope Slides, Epredia
Fournisseur: Thermo Scientific
Ward's® Luminol Powder Concentrate
Fournisseur: EVIDENT CRIME SCENE PRODUCTS
Histoplex™ Histology Containers, Starplex®
Fournisseur: Starplex
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Papanicolaou's Orange G solution (OG 6), VWR® stain for histology, for cytology
Fournisseur: VWR International
Imidazole 99%
Fournisseur: Thermo Scientific Chemicals
Plasmid DNA Buffers, Omega Bio-tek
Fournisseur: Omega Bio-Tek
Horse Red Blood Cells 10% Washed Pooled Cells, Rockland Immunochemicals
Fournisseur: Rockland Immunochemical
DPD Free Chlorine Reagent Powder Pillows, 5 ml, Hach
Fournisseur: Hach
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Fournisseur: Sekisui Diagnostics
Acetonedimethyl acetal 98%
Fournisseur: Thermo Scientific Chemicals
Polysorbate 80 NF, Multi-Compendial, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC