PathFlow Supa Mega Cassettes | Avantor
PathFlow Supa Mega Cassettes
PathFlow Supa Mega cassettes, Pink
Catalog # 77908-008
New Product
Supplier:   StatLab
Fournisseur #:  EAG-0108-02A
PathFlow Supa Mega Cassettes
Catalog # 77908-008
Supplier:  StatLab
New Product
PathFlow Supa Mega Cassettes
PathFlow Supa Mega cassettes, Pink
Price
$619.95
Log in pour la disponibilité
Box of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Supa Mega Cassette is particularly useful for prostate, brain, breast tissue, and eye. As a result, this cassette has been widely adopted by clinical, research, and toxicology laboratories.
  • Better reagent flow and improved turnaround times
    Proven writing surface
    Large color range available
The Supa Mega Cassette design offers better sectioning quality, with the innovative hex design ensuring a greater adhesion of the paraffin block to the cassette. This eliminates the possibility of the embedded specimen being separated from the cassette when sectioning fibrous specimens.
Spécifications
  • Color:
    Pink
  • Désignation:
    PathFlow Supa Mega cassettes