PathFlow Supa Mega Cassettes
PathFlow Supa Mega cassettes, Grey
  77908-004
New Product
 :   StatLab
 :  EAG-0105-02A
PathFlow Supa Mega Cassettes
  77908-004
 :  StatLab
New Product
PathFlow Supa Mega Cassettes
PathFlow Supa Mega cassettes, Grey
$619.95
Box of 100
The Supa Mega Cassette is particularly useful for prostate, brain, breast tissue, and eye. As a result, this cassette has been widely adopted by clinical, research, and toxicology laboratories.
  • Better reagent flow and improved turnaround times
    Proven writing surface
    Large color range available
The Supa Mega Cassette design offers better sectioning quality, with the innovative hex design ensuring a greater adhesion of the paraffin block to the cassette. This eliminates the possibility of the embedded specimen being separated from the cassette when sectioning fibrous specimens.
  • Color:
    Grey
  • Désignation:
    PathFlow Supa Mega cassettes