Slide Boxes, HDPE
Blue plastic slide boxes made from durable HDPE .
- Hinged lid
- Foam interior cushioning
- Equipped with locking mechanism
- 12 boxes in each case
- Color:Blue
- Désignation:50-place slide box
- Matière:HDPE