PathFlow Supa Mega Slim Cassettes
PathFlow supa mega slim cassettes, Blue
  77908-018
New Product
PathFlow Supa Mega Slim Cassettes
  77908-018
New Product
PathFlow Supa Mega Slim Cassettes
PathFlow supa mega slim cassettes, Blue
$420.63
Box of 100
The Supa Mega Slim cassette is being widely adopted in clinical, research, and toxicology laboratories.
  • Casting can be done in 20 minutes at −8 °C
  • Large color range available
  • Specific base molds
The Supa Mega Slim cassette offers better reagent flow and improved turnaround times, with less wax used during the embedding process.
  • Color:
    Blue
  • Désignation:
    PathFlow supa mega slim cassettes