PathFlow Supa Mega Slim Cassettes
Product Details & Documents
The Supa Mega Slim cassette is being widely adopted in clinical, research, and toxicology laboratories.
- Casting can be done in 20 minutes at −8 °C
- Large color range available
- Specific base molds
The Supa Mega Slim cassette offers better reagent flow and improved turnaround times, with less wax used during the embedding process.
Spécifications
- Color:Green
- Désignation:PathFlow supa mega slim cassettes